R&D Systems
Manufacturer Code:FAB14001C100
Catalog # FAB14001C10
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 218213 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human IL-23 R Gly24-Ile354 Accession # Q5VWK5 |
Conjugate | PerCP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: IBD17 IL-23 R IL-23 receptor IL23R IL-23R interleukin 23 receptor interleukin-23 receptor; IL-23 receptor; Interleukin-23 receptor
Gene Aliases: IL23R
UniProt ID: (Human) Q5VWK5
Entrez Gene ID: (Human) 149233
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.