R&D Systems
Manufacturer Code:MAB2173100
Catalog # MAB2173100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 948536 |
Immunogen | <em>S. frugiperda</em> insect ovarian cell line <em>Sf</em> 21-derived recombinant human IL-10 Met1-Asn178 Accession # P22301 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CSIF; CSIF CSIFMGC126450 Cytokine synthesis inhibitory factor GVHDS IL10 IL10A IL-10MGC126451 interleukin 10 interleukin-10 TGIF; Cytokine synthesis inhibitory factor; IL-10; Interleukin-10; T-cell growth inhibitory factor
Gene Aliases: CSIF; GVHDS; IL-10; IL10; IL10A; TGIF
UniProt ID: (Human) P22301
Entrez Gene ID: (Human) 3586
Molecular Function: cytokine interleukin superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.