Invitrogen
Manufacturer Code:PA569977
Catalog # PIPA569977
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human IFT88 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IFT88 polaris TTC10 tetratricopeptide repeat domain 10 HGNC:20606 D13S1056E DAF19 IFT88 MGC26259 RP11-172H24.2 TG737 hTg737 TPR repeat protein 10 Probe hTg737 (polycystic kidney disease autosomal recessive) Tg737 protein intraflagellar transport 88 homolog recessive polycystic kidney disease protein Tg737 homolog intraflagellar transport 88 homolog (Chlamydomonas); intraflagellar transport 88 homolog; intraflagellar transport protein; intraflagellar transport protein 88; Intraflagellar transport protein 88 homolog; oval; ovl; polaris; polaris homolog; polaris; axonemal transport protein; probe hTg737 (polycystic kidney disease, autosomal recessive); Recessive polycystic kidney disease protein Tg737; Recessive polycystic kidney disease protein Tg737 homolog; testicular tissue protein Li 93; tetratricopeptide repeat domain 10; Tetratricopeptide repeat protein 10; TgN(Imorpk)737Rpw; TPR repeat protein 10; transgene insert site 737, insertional mutation, polycystic kidney disease
Gene Aliases: AW552028; D13S1056E; DAF19; fb37b11; flexo; fxo; hTg737; IFT88; orpk; osm-5; polaris; TG737; Tg737Rpw; TgN737Rpw; TTC10; wu:fb37b11; xx:zah8127
UniProt ID: (Human) Q13099, (Mouse) Q61371
Entrez Gene ID: (Human) 8100, (Bovine) 514177, (Rabbit) 100009576, (Dog) 477345, (Horse) 100054328, (Mouse) 21821, (Guinea pig) 100714887, (Rat) 305918, (Zebrafish) 321855
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.