Invitrogen
Manufacturer Code:MA530506
Catalog # PIMA530506
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 126 |
Immunogen | Recombit Human IFNA5/IFNaG Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: IFN-alpha-5; Interferon alpha-5; Interferon alpha-5 IFN-alpha-5 Interferon alpha-61 Interferon alpha-G LeIF G IFNA5; Interferon alpha-61; Interferon alpha-G; interferon, alpha 5; LeIF G
Gene Aliases: IFN-alpha-5; IFN-alphaG; IFNA5; INA5; INFA5; leIF G
UniProt ID: (Human) P01569
Entrez Gene ID: (Human) 3442
Molecular Function: cytokine interferon superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.