Abnova
Manufacturer Code:PAB25302
Catalog # 89-126-961
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding Y512 of human ICAM1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ICAM1.
For Research Use Only
Protein Aliases: BB2; CD54; cell surface glycoprotein P3.58; ICAM-1; Intercellular adhesion molecule 1; intercellular adhesion molecule 1 (CD54), human rhinovirus receptor; Major group rhinovirus receptor; P3.58
Gene Aliases: BB2; CD54; ICAM1; P3.58
UniProt ID: (Human) P05362
Entrez Gene ID: (Human) 3383
Molecular Function: cell adhesion molecule immunoglobulin superfamily cell adhesion molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.