Novus Biologicals
Manufacturer Code:NBP270891JF646
Catalog # N270891J646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4C5 |
Immunogen | Full length recombinant protein of human Hexokinase 2 (NP_000180) produced in HEK293T cells. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: DKFZp686M1669 EC 2.7.1 hexokinase 2 Hexokinase type II HKII HXK2 muscle Muscle form hexokinase; Hexokinase type II; Hexokinase-2; hexokinase-2, muscle; Hexokinase-B; HK II; Muscle form hexokinase
Gene Aliases: HK2; HKII; HXK2
UniProt ID: (Human) P52789
Entrez Gene ID: (Human) 3099
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.