Invitrogen
Manufacturer Code:PA553076
Catalog # PIPA553076
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human HSPA12B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Heat shock 70 kDa protein 12B; Heat shock 70 kDa protein 12B HSPA12B C20orf60; heat shock protein 70kDa 12B
Gene Aliases: 2700081N06Rik; C20orf60; HSPA12B
UniProt ID: (Human) Q96MM6, (Mouse) Q9CZJ2
Entrez Gene ID: (Human) 116835, (Rat) 311427, (Mouse) 72630
Molecular Function: Hsp70 family chaperone chaperone kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.