Invitrogen
Manufacturer Code:PA5100821
Catalog # PIPA5100821
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human HSPA12B corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2700081N06Rik C20orf60 Heat shock 70 kDa protein 12B Heat shock protein heat shock protein 12B heat shock protein 70kDa 12B heat shock protein family A (Hsp70) member 12B HSP Hspa12b; Heat shock 70 kDa protein 12B
Gene Aliases: 2700081N06Rik; C20orf60; HSPA12B
UniProt ID: (Human) Q96MM6, (Mouse) Q9CZJ2
Entrez Gene ID: (Human) 116835, (Mouse) 72630
Molecular Function: Hsp70 family chaperone chaperone kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.