Invitrogen
Manufacturer Code:PA577793
Catalog # PIPA577793
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide derived from the sequence of hamster HSP110 sequence identical to human and mouse |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 42 degrees C-HSP; heat shock 105kDa; Heat shock 105kDa alpha heat shock 105kDa beta HSP105 HSP105A HSP105B HSP110 HSPH1 NY CO 25; heat shock 105kDa/110kDa protein 1; Heat shock 110 kDa protein; Heat shock protein 105 kDa; heat shock protein 110; heat shock protein, 105 kDa; Heat shock-related 100 kDa protein E7I; heat-shock protein 110 kDa; HSP-E7I; HSP105 42 C-HSP; Septin-8
Gene Aliases: 105kDa; AI790491; hsp-E7I; HSP105; HSP110; hsp110/105; HSPH1; I79_002075; Kiaa0201; KIAA0202; SEP2; SEPT8; SEPTIN8
UniProt ID: (Mouse) Q61699, (Rat) Q66HA8, (Chinese hamster) Q60446, (Human) Q92599, (Bovine) Q0VCP4
Entrez Gene ID: (Mouse) 15505, (Rat) 288444, (Chinese hamster) 100689462, (Human) 23176, (Rhesus monkey) 708424, (Bovine) 540614, (Sheep) 101122277
Molecular Function: G-protein cytoskeletal protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.