Novus Biologicals
Manufacturer Code:NBP19132420UL
Catalog # NBP19132420
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C terminal of human HS3ST6. Peptide sequence GERLVSDPAGEVGRVQDFLGLKRVVTDKHFYFNATKGFPCLKKAQGGSRP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-OST-6; h3-OST-6 heparan sulfate (glucosamine) 3-O-sulfotransferase 6 heparan sulfate 3-O-sulfotransferase 6 heparan sulfate glucosamine 3-O-sulfotransferase 6 HS3ST5; heparan sulfate (glucosamine) 3-O-sulfotransferase 5; heparan sulfate (glucosamine) 3-O-sulfotransferase 6; Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 6; Heparan sulfate glucosamine 3-O-sulfotransferase 6; heparan sulphate D-glucosaminyl 3-O-sulfotransferase-3B like
Gene Aliases: 3-OST-6; HS3ST5; HS3ST6
UniProt ID: (Human) Q96QI5
Entrez Gene ID: (Human) 64711
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.