Invitrogen
Manufacturer Code:PA595381
Catalog # PIPA595381
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human HPRT recombit protein (Position: A2-S110). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HGPRT; HGPRTase; HPRT B; hypoxanthine guanine phosphoribosyl transferase 1; hypoxanthine phosphoribosyl transferase; Hypoxanthine-guanine phosphoribosyltransferase; hypoxanthine-guanine phosphoribosyltransferase 1; Hypoxanthine-guanine phosphoribosyltransferase HGPRT HGPRTase 2.4.2.8 HPRT1 HPRT; testicular tissue protein Li 89
Gene Aliases: C81579; HGPRT; Hgprtase; HPGRT; HPRT; HPRT1
UniProt ID: (Human) P00492, (Mouse) P00493, (Rat) P27605
Entrez Gene ID: (Human) 3251, (Mouse) 15452, (Rat) 24465
Molecular Function: glycosyltransferase isomerase mutase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.