Abnova
Manufacturer Code:H00081502K
Catalog # 89-209-462
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human HM13 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human HM13 peptide using ARM Technology.
For Research Use Only
Protein Aliases: H13 IMP1 IMPAS MSTP086 PSENL3 PSL3 SPP dJ324O17.1; histocompatibility (minor) 13; histocompatibility 13; IMP-1; Intramembrane protease 1; minor histocompatibility antigen 13; Minor histocompatibility antigen H13; Presenilin-like protein 3; Signal peptide peptidase; signal peptide peptidase beta; signal peptide peptidase like 1
Gene Aliases: H13; HM13; IMP1; IMPAS; IMPAS-1; MSTP086; PSENL3; PSL3; SPP; SPPL1
UniProt ID: (Human) Q8TCT9
Entrez Gene ID: (Human) 81502
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.