Invitrogen
Manufacturer Code:PA588165
Catalog # PIPA588165
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 178-397 of human DNAJA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DnaJ (Hsp40) homolog, subfamily A, member 1; DnaJ homolog subfamily A member 1; DnaJ protein homolog 2; DnaJ-like protein 1; DnaJ-like protein 2; hDj-2; Heat shock 40 kDa protein 4; Heat shock protein J2; heat shock protein, DNAJ-like 2; HSDJ; HSJ-2; Human DnaJ protein 2; neural precursor cell expressed, developmentally down-regulated 7; neural precursor cell expressed, developmentally down-regulated gene 7
Gene Aliases: DJ-2; DjA1; DNAJ2; DNAJA1; hDJ-2; HDJ2; HSDJ; HSJ-2; HSJ2; HSPF4; NEDD7; Rdj1
UniProt ID: (Human) P31689, (Mouse) P63037, (Rat) P63036
Entrez Gene ID: (Human) 3301, (Mouse) 15502, (Rat) 65028
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.