Invitrogen
Manufacturer Code:PA572486
Catalog # PIPA572486
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 737-763 amino acids from the C-terminal region of human HADHA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-ketoacyl-Coenzyme A (CoA) thiolase, alpha subunit; 3-oxoacyl-CoA thiolase; 78 kDa gastrin-binding protein; ECHA GBP HADH LCEH LCHAD MTPA TP-ALPHA 3-ketoacyl-Coenzyme A (CoA) thiolase alpha subunit 3-oxoacyl-CoA thiolase 78 kDa gastrin-binding protein gastrin-binding protein hydroxyacyl-Coenzyme A dehydrogenase/3-ketoacyl-Coenzyme A thiolase/enoyl-Coenzyme A hydratase (trifunctional protein) alpha; gastrin-binding protein; hydroxyacyl-Coenzyme A dehydrogenase/3-ketoacyl-Coenzyme A thiolase/enoyl-Coenzyme A hydratase (trifunctional protein), alpha subunit; Long chain 3-hydroxyacyl-CoA dehydrogenase; long-chain 2-enoyl-CoA hydratase; Long-chain enoyl-CoA hydratase; long-chain-3-hydroxyacyl-CoA dehydrogenase; mitochondrial long-chain 2-enoyl-Coenzyme A (CoA) hydratase, alpha subunit; mitochondrial long-chain L-3-hydroxyacyl-Coenzyme A (CoA) dehydrogenase, alpha subunit; mitochondrial trifunctional enzyme, alpha subunit; mitochondrial trifunctional protein, alpha subunit; Monolysocardiolipin acyltransferase; TP-alpha; Trifunctional enzyme subunit alpha, mitochondrial
Gene Aliases: ECHA; GBP; HADH; HADHA; LCEH; LCHAD; MTPA; TP-ALPHA
UniProt ID: (Human) P40939
Entrez Gene ID: (Human) 3030
Molecular Function: dehydrogenase epimerase/racemase hydratase isomerase lyase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.