Invitrogen
Manufacturer Code:PA519654
Catalog # PIPA519654
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH derived from within residues 400 - 500 of Human H Cadherin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cadherin 13, H-cadherin (heart); Cadherin-13; CDHH H-cadherin P105 T-cad T-cadherin cadherin-13 heart cadherin heart-cadherin truncated cadherin truncated-cadherin; H-cadherin; H-cadherin (heart); Heart cadherin; P105; T-cad; T-cadherin; Truncated cadherin
Gene Aliases: 4932416G01Rik; CDH13; CDHH; Cdht; P105; T-cadherin; Tcad
UniProt ID: (Human) P55290, (Mouse) Q9WTR5
Entrez Gene ID: (Human) 1012, (Mouse) 12554
Molecular Function: cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.