Novus Biologicals
Manufacturer Code:NBP290374JF549
Catalog # NB038999
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 8 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Mouse Growth Hormone R (NP_034414.2 Met1-Gln273). |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: GH receptor; GH receptor GHBP growth hormone binding protein growth hormone receptor serum binding protein Somatotropin receptor; GH-binding protein; growth hormone binding protein; Growth hormone receptor; Growth hormone-binding protein; serum binding protein; Serum-binding protein; Somatotropin receptor
Gene Aliases: GHBP; GHIP; GHR
UniProt ID: (Human) P10912
Entrez Gene ID: (Human) 2690
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.