Novus Biologicals
Manufacturer Code:NBP220017
Catalog # NBP220017
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of human GPBB. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Brain glycogen phosphorylase EC 2.4.1.1 Glycogen phosphorylase B Glycogen phosphorylase brain form Glycogen Phosphorylase Isoenzyme BB MGC9213 Phosphorylase glycogen brain PYGB; glycogen phosphorylase B; Glycogen phosphorylase, brain form
Gene Aliases: GPBB; PYGB
UniProt ID: (Human) P11216
Entrez Gene ID: (Human) 5834
Molecular Function: phosphorylase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.