Novus Biologicals
Manufacturer Code:NBP27971120UG
Catalog # NB010440
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CPTC-GPI-1 |
Immunogen | Recombinant human full-length Glucose 6 phosphate isomerase protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AMF; AMFGNPI Autocrine motility factor DKFZp686C13233 EC 5.3.1.9 glucose phosphate isomerase glucose-6-phosphate isomerase hexose monophosphate isomerase hexosephosphate isomerase neuroleukin NLKSA36 oxoisomerase PGI PHI Phosphoglucose isomerase phosphohexomutase Phosphohexose isomerase phosphosaccharomutase SA-36 Sperm antigen 36 sperm antigen-36; Autocrine motility factor; glucose phosphate isomerase; Glucose-6-phosphate isomerase; GPI; hexose monophosphate isomerase; hexosephosphate isomerase; Neuroleukin; NLK; oxoisomerase; PGI; PHI; Phosphoglucose isomerase; phosphohexomutase; Phosphohexose isomerase; phosphosaccharomutase; SA-36; Sperm antigen 36; sperm antigen-36
Gene Aliases: AMF; GNPI; GPI; NLK; PGI; PHI; SA-36; SA36
UniProt ID: (Human) P06744
Entrez Gene ID: (Human) 2821
Molecular Function: isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.