Invitrogen
Manufacturer Code:PA520660
Catalog # PIPA520660
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 15 amino acid peptide near the carboxy terminus of human Gle1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GLE1 RNA export mediator homolog; GLE1 RNA export mediator-like (yeast; GLE1-like protein; GLE1-like, RNA export mediator; GLE1L LCCS LCCS1 hGLE1 GLE1-like protein GLE1-like RNA export mediator nucleoporin GLE1; hGLE1; Nucleoporin GLE1
Gene Aliases: 4933405K21Rik; AA553313; GLE1; GLE1L; hGLE1; LCCS; LCCS1
UniProt ID: (Human) Q53GS7, (Mouse) Q8R322, (Rat) Q4KLN4
Entrez Gene ID: (Human) 2733, (Mouse) 74412, (Rat) 362098
Molecular Function: nucleic acid binding transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.