Abnova
Manufacturer Code:PAB11663
Catalog # 89-115-269
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of rat Gclc. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Rabbit polyclonal antibody raised against synthetic peptide of Gclc.
For Research Use Only
Protein Aliases: Gamma-ECS; Gamma-glutamylcysteine synthetase; GCS heavy chain; Glclc; Glutamate--cysteine ligase catalytic subunit; Glutamylcysteine gamma synthetase light chain; MGC93096
Gene Aliases: Gclc; Glclc
UniProt ID: (Rat) P19468
Entrez Gene ID: (Rat) 25283
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.