Proteintech
Manufacturer Code:223711AP150UL
Catalog # 50-173-3850
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GYS2 Fusion Protein Ag18012 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glycogen [starch] synthase, liver; glycogen synthase 2 (liver); glycogen synthase 2 (liver) GYS2; glycogen synthase, liver
Gene Aliases: BC021322; GLYSN; GYS2; LGS
UniProt ID: (Human) P54840, (Mouse) Q8VCB3
Entrez Gene ID: (Human) 2998, (Mouse) 232493, (Rat) 25623
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.