Novus Biologicals
Manufacturer Code:NBP212342APCCY7
Catalog # N212342APC7
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0316-8W1 |
Immunogen | Human GM-CSF Receptor alpha |
Conjugate | APC-Cyanine7 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: CD116; CD116 antigen; CD116 CD116 antigen CDw116 colony stimulating factor 2 receptor alpha subunit colony stimulating factor 2 receptor alpha low-affinity(granulocyte-macrophage) CSF2R CSF2RAX CSF2RAY CSF2RX CSF2RY GM-CSF receptor alpha subunit GMCSFR GM-CSF-R-alpha GMCSFR-alpha GMR GMR-alpha granulocyte-macrophage colony-stimulating factor receptor alpha chain granulocyte-macrophage colony-stimulating factor receptor subunit alpha MGC3848 MGC4838; CDw116; colony stimulating factor 2 receptor, alpha, low-affinity (granulocyte-macrophage); GM-CSF receptor alpha subunit; GM-CSF-R-alpha; GMCSFR-alpha; GMR-alpha; granulocyte-macrophage colony-stimulating factor receptor alpha chain; Granulocyte-macrophage colony-stimulating factor receptor subunit alpha
Gene Aliases: CD116; CDw116; CSF2R; CSF2RA; CSF2RAX; CSF2RAY; CSF2RX; CSF2RY; GM-CSF-R-alpha; GMCSFR; GMR; SMDP4
UniProt ID: (Human) P15509
Entrez Gene ID: (Human) 1438
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.