Abnova
Manufacturer Code:H00002733M03
Catalog # 89-022-320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D8 |
Immunogen | GLE1 (NP_001003722.1 140 a.a. ~ 240 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant GLE1.
For Research Use Only
Protein Aliases: GLE1 RNA export mediator homolog; GLE1-like protein; GLE1-like, RNA export mediator; GLE1L; hGLE1; LCCS; LCCS1; Nucleoporin GLE1
Gene Aliases: GLE1; GLE1L; hGLE1; LCCS; LCCS1
UniProt ID: (Human) Q53GS7
Entrez Gene ID: (Human) 2733
Molecular Function: nucleic acid binding transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.