Novus Biologicals
Manufacturer Code:NBP132714
Catalog # NBP132714
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human GLE1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GLE1 (yeast homolog)-like RNA export mediator GLE1 RNA export mediator homolog (yeast) GLE1LGLE1 RNA export mediator-like (yeast) GLE1-like protein GLE1-like RNA export mediator hGLE1GLE1 RNA export mediator (yeast) LCCS LCCS1 lethal congenital contracture syndrome 1 nucleoporin GLE1; GLE1 RNA export mediator homolog; GLE1-like protein; GLE1-like, RNA export mediator; hGLE1; Nucleoporin GLE1
Gene Aliases: GLE1; GLE1L; hGLE1; LCCS; LCCS1
UniProt ID: (Human) Q53GS7
Entrez Gene ID: (Human) 2733
Molecular Function: nucleic acid binding transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.