Invitrogen
Manufacturer Code:PA546853
Catalog # PIPA546853
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human GCM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Chorion-specific transcription factor GCMa; Gcm 1; Gcm a; GCM motif protein 1; glial cell deficient; Glial cells missing homolog 1; Glial cells missing homolog 1 (Drosophila) GCMA hGCMa; hGCMa; mGCM1; mGCMa
Gene Aliases: Gcm-rs2; GCM1; Gcm1-rs1; GCMA; glide; hGCMa
UniProt ID: (Human) Q9NP62, (Rat) Q9Z288, (Mouse) P70348
Entrez Gene ID: (Dog) 481844, (Human) 8521, (Rat) 29394, (Mouse) 14531
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.