Invitrogen
Manufacturer Code:PA569764
Catalog # PIPA569764
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of rat FXN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: d-FXN; DKEYP-118A9.1; Frataxin intermediate form; Frataxin mature form; Frataxin(56-210); Frataxin(78-210); Frataxin(81-210); Frataxin, mitochondrial; Friedreich ataxia; Friedreich ataxia protein; Friedreich ataxia protein frataxin mitochondrial Frataxin mitochondrial Frataxin intermediate form FRDA X25; Fxn; i-FXN; m56-FXN; m78-FXN; m81-FXN
Gene Aliases: CyaY; FA; FARR; FRDA; FXN; RGD1565754; X25; zgc:158218
UniProt ID: (Human) Q16595, (Bovine) Q05B87, (Mouse) O35943
Entrez Gene ID: (Human) 2395, (Bovine) 505694, (Horse) 100056887, (Dog) 609051, (Rabbit) 103348216, (Rat) 499335, (Guinea pig) 100729273, (Mouse) 14297, (Zebrafish) 556389
Molecular Function: cation transporter kinase transferase transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.