Proteintech
Manufacturer Code:127641AP150UL
Catalog # 50-172-8233
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FOXG1 Fusion Protein Ag3426 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BF 1 BF 2 BF1 BF2 Brain factor 1 Brain factor 2 FHKL3 FKH2 FKHL1 FKHL2 FKHL3 FKHL4 forkhead box G1 Forkhead box protein G1 Forkhead box protein G1A Forkhead box protein G1B Forkhead box protein G1C Forkhead related protein FKHL1 Forkhead related protein FKHL2 Forkhead related protein FKHL3 FOXG1 FOXG1A FOXG1B FOXG1C HBF 1 HBF 2 HBF 3 HBF G2 HBF2 HFK1 HFK2 HFK3 KHL2 QIN; BF-1; BF-2; BF1; Brain factor 1; Brain factor 2; Forkhead box protein G1; Forkhead box protein G1A; Forkhead box protein G1B; Forkhead box protein G1C; forkhead-like 1; forkhead-like 2; forkhead-like 3; forkhead-like 4; forkhead-like transcription factor BF-1; Forkhead-related protein FKHL1; Forkhead-related protein FKHL2; Forkhead-related protein FKHL3; FoxG1; HFK1; HFK2; HFK3; HNF-3/forkhead homolog, brain factor 1; oncogene QIN; transcription factor BF-1
Gene Aliases: 2900064B05Rik; BF-1; BF1; BF1A; BF2; FHKL3; FKH2; FKHL1; FKHL2; FKHL3; FKHL4; FOXG1; FOXG1A; FOXG1B; FOXG1C; HBF-1; HBF-2; HBF-3; HBF-G2; HBF2; Hfh9; Hfhbf1; HFK1; HFK2; HFK3; KHL2; QIN; RATBF1A
UniProt ID: (Human) P55316, (Rat) Q00939, (Mouse) Q60987
Entrez Gene ID: (Human) 2290, (Rat) 24370, (Mouse) 15228
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.