Invitrogen
Manufacturer Code:PA570059
Catalog # PIPA570059
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human FLVCR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: eline leukemia virus subgroup C cellular receptor 1; feline leukemia virus subgroup C cellular receptor 1; Feline leukemia virus subgroup C receptor; Feline leukemia virus subgroup C receptor-related protein 1; FLVCR1 feline leukemia virus subgroup C cellular receptor FLVCR protein FLVCR eline leukemia virus subgroup C cellular receptor 1; Major facilitator superfamily domain containing 7B; Mfsd7b
Gene Aliases: 9630055N22Rik; AXPC1; FLVCR; FLVCR1; MFSD7B; PCA; PCARP
UniProt ID: (Human) Q9Y5Y0, (Mouse) B2RXV4
Entrez Gene ID: (Human) 28982, (Bovine) 533317, (Horse) 100050536, (Dog) 100856001, (Rabbit) 100358453, (Guinea pig) 100720993, (Rat) 683955, (Mouse) 226844
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.