Novus Biologicals
Manufacturer Code:NBP299396B
Catalog # NB083460
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant FGF-10 (NP_004456.1 Gln38-Ser208). |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FGF-10; FGF-10 fibroblast growth factor 10 Keratinocyte growth factor 2 produced by fibroblasts of urinary bladder lamina propria; Fibroblast growth factor 10; Keratinocyte growth factor 2; produced by fibroblasts of urinary bladder lamina propria
Gene Aliases: FGF10
UniProt ID: (Human) O15520
Entrez Gene ID: (Human) 2255
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.