Novus Biologicals
Manufacturer Code:NBP290812H
Catalog # NB078314
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 115 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Mouse FDPS (NP_608219.1 Met1-Lys353). |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: (2E,6E)-farnesyl diphosphate synthase; 6E)-farnesyl diphosphate synthase Dimethylallyltranstransferase EC 2.5.1.1 EC 2.5.1.10 farnesyl diphosphate synthaseFPP synthetase farnesyl pyrophosphate synthase farnesyl pyrophosphate synthetase dimethylallyltranstransferase FPP synthase FPPS FPSgeranyltranstransferase Geranyltranstransferase KIAA1293; Dimethylallyltranstransferase; Farnesyl diphosphate synthase; Farnesyl pyrophosphate synthase; farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, geranyltranstransferase; FPP synthase; FPP synthetase; Geranyltranstransferase
Gene Aliases: FDPS; FPPS; FPS; KIAA1293; POROK9
UniProt ID: (Human) P14324
Entrez Gene ID: (Human) 2224
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.