Invitrogen
Manufacturer Code:MA534738
Catalog # PIMA534738
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JG35-83 |
Immunogen | Recombinant protein within human Emi1 aa 50-250. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 2510044I10Rik C85305 Early mitotic inhibitor 1 EMI1 F-box only protein 5 FBX5 Fbxo31 Fbxo5; Early mitotic inhibitor 1; F-box only protein 5; F-box protein Fbx5
Gene Aliases: EMI1; FBX5; Fbxo31; FBXO5
UniProt ID: (Human) Q9UKT4
Entrez Gene ID: (Human) 26271
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.