Novus Biologicals
Manufacturer Code:NBP237738
Catalog # NBP237738
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6B3A9 |
Immunogen | Purified recombinant fragment of human SLC27A2 (AA: 346-405) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ACSVL1FATP-2 EC 6.2.1 EC 6.2.1.- EC 6.2.1.3 EC 6.2.1.7 FACVL1very long-chain fatty-acid-coenzyme A ligase 1 FATP2THCA-CoA ligase Fatty acid transport protein 2 Fatty-acid-coenzyme A ligase very long-chain 1 hFACVL1 HsT17226 Long-chain-fatty-acid--CoA ligase solute carrier family 27 (fatty acid transporter) member 2 Solute carrier family 27 member 2 very-long-chain acyl-CoA synthetase VLACSVery long-chain-fatty-acid-CoA ligase VLCSvery long-chain acyl-CoA synthetase; Arachidonate--CoA ligase; FATP-2; Fatty acid transport protein 2; Fatty-acid-coenzyme A ligase, very long-chain 1; Long-chain-fatty-acid--CoA ligase; Phytanate--CoA ligase; solute carrier family 27 (fatty acid transporter), member 2; Solute carrier family 27 member 2; THCA-CoA ligase; Very long-chain acyl-CoA synthetase; very long-chain fatty-acid-coenzyme A ligase 1; Very long-chain-fatty-acid-CoA ligase; very-long-chain acyl-CoA synthetase; VLACS
Gene Aliases: ACSVL1; FACVL1; FATP2; hFACVL1; HsT17226; SLC27A2; VLACS; VLCS
UniProt ID: (Human) O14975
Entrez Gene ID: (Human) 11001
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.