Novus Biologicals
Manufacturer Code:NBP304536100UL
Catalog # NB094360
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human FAR1 (NP_115604.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: fatty acyl CoA reductase 1; fatty acyl CoA reductase 1 FLJ22728 FLJ33561 short chain dehydrogenase/reductase family 10E member 1 UNQ2423/PRO4981; Fatty acyl-CoA reductase 1; Male sterility domain-containing protein 2; short chain dehydrogenase/reductase family 10E, member 1
Gene Aliases: FAR1; MLSTD2; PFCRD; SDR10E1; UNQ2423/PRO4981
UniProt ID: (Human) Q8WVX9
Entrez Gene ID: (Human) 84188
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.