Invitrogen
Manufacturer Code:PA530080
Catalog # PIPA530080
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 247 and 444 of Human FADS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA (8-3)-desaturase; D5D; D5D FADS6 FADSD5 LLCDL1 TU12 delta(5) desaturase delta(5) fatty acid desaturase delta-5 desaturase delta-5 fatty acid desaturase linoleoyl-CoA desaturase (delta-6-desaturase)-like 1; delta(5) desaturase; Delta(5) fatty acid desaturase; delta-5 desaturase; delta-5 fatty acid desaturase; Fatty acid desaturase 1; linoleoyl-CoA desaturase (delta-6-desaturase)-like 1
Gene Aliases: D5D; FADS1; FADS6; FADSD5; LLCDL1; TU12
UniProt ID: (Human) O60427
Entrez Gene ID: (Human) 3992
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.