OriGene
Manufacturer Code:CF805454
Catalog # 50-166-7435
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2F11 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-252 of human ESRRB produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DFNB35 ERR2 ERRb ESRL2 NR3B2; ERR beta-2; ERR-beta; ERRbeta-2; Estrogen receptor-like 2; estrogen-related nuclear receptor beta; Estrogen-related receptor beta; nuclear receptor ERRB2; Nuclear receptor subfamily 3 group B member 2; orphan nuclear receptor; Steroid hormone receptor ERR2
Gene Aliases: DFNB35; ERR2; ERRb; ERRB2; ESRL2; ESRRB; NR3B2
UniProt ID: (Human) O95718
Entrez Gene ID: (Human) 2103
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.