Abnova
Manufacturer Code:PAB25698
Catalog # 89-127-320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 19 amino acids at C-terminus of human ESRRB. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ESRRB.
For Research Use Only
Protein Aliases: DFNB35; ERR beta-2; ERR-beta; ERR2; ERRb; ERRbeta; ERRbeta-2; ESRL2; Estrogen receptor-like 2; estrogen-related nuclear receptor beta; Estrogen-related receptor beta; NR3B2; nuclear receptor ERRB2; Nuclear receptor subfamily 3 group B member 2; orphan nuclear receptor; Steroid hormone receptor ERR2
Gene Aliases: DFNB35; ERR2; ERRb; ERRB2; ESRL2; ESRRB; NR3B2
UniProt ID: (Human) O95718
Entrez Gene ID: (Human) 2103
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.