Novus Biologicals
Manufacturer Code:NBP232344
Catalog # NBP232344
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This ESD antibody is generated from a rabbit immunized with a KLH conjugated synthetic peptide between 68-102 amino acids from the Central region of human ESD. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.2.12 esterase 10 esterase Desterase D/formylglutathione hydrolase FGH FLJ11763 S-formylglutathione hydrolase; esterase 10; Esterase D; esterase D/formylglutathione hydrolase; FGH; Methylumbelliferyl-acetate deacetylase; S-formylglutathione hydrolase; testicular tissue protein Li 66
Gene Aliases: ESD; FGH
UniProt ID: (Human) P10768
Entrez Gene ID: (Human) 2098
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.