Invitrogen
Manufacturer Code:PA522745
Catalog # PIPA522745
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made the the N-terminal region of the human protein. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Endoplasmic oxidoreductin-1-like protein; endoplasmic reticulum oxidation 1; Endoplasmic reticulum oxidoreductase alpha; Endoplasmic reticulum oxidoreductin-1-like protein; ERO1-L; ERO1-L-alpha; ERO1-like protein alpha; Global ischemia-induced protein 11; Oxidoreductin-1-L-alpha; UNQ434/PRO865 ERO1-alpha ERO1A ERO1-L ERO1-L-alpha ERO1-like protein alpha endoplasmic oxidoreductin-1-like protein oxidoreductin-1-L-alpha
Gene Aliases: ERO1-alpha; ERO1-L; ERO1-L-alpha; ERO1A; Ero1alpha; ERO1L; ERO1LA; Giig11; UNQ434/PRO865
UniProt ID: (Human) Q96HE7, (Mouse) Q8R180, (Rat) Q8R4A1
Entrez Gene ID: (Human) 30001, (Mouse) 50527, (Rat) 171562
Molecular Function: oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.