Novus Biologicals
Manufacturer Code:NB1002525AF700100UL
Catalog # NB003454
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to the N-terminal region of human ERO1L [UniProt Q96HE7] |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 1.8.4 EC 1.8.4.- Endoplasmic oxidoreductin-1-like protein ERO1 (S. cerevisiae)-like ERO1A ERO1-alpha ERO1-L ERO1-L-alpha ERO1-like (S. cerevisiae) ERO1-like protein alpha oxidoreductin-1-L-alpha; Endoplasmic oxidoreductin-1-like protein; Endoplasmic reticulum oxidoreductase alpha; ERO1-L; ERO1-like protein alpha; Oxidoreductin-1-L-alpha
Gene Aliases: ERO1-alpha; ERO1-L; ERO1-L-alpha; ERO1A; Ero1alpha; ERO1L; ERO1LA; UNQ434/PRO865
UniProt ID: (Human) Q96HE7
Entrez Gene ID: (Human) 30001
Molecular Function: oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.