Abnova
Manufacturer Code:H00002027M01A
Catalog # 89-104-440
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5D1 |
Immunogen | ENO3 (NP_001967228 a.a. &bsim 277 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant ENO3.
For Research Use Only
Protein Aliases: 2-phospho-D-glycerate hydro-lyase; 2-phospho-D-glycerate hydrolyase; Beta-enolase; Enolase 3; enolase 3 (beta, muscle); MSE; muscle enriched enolase; Muscle-specific enolase; Skeletal muscle enolase
Gene Aliases: ENO3; GSD13; MSE
UniProt ID: (Human) P13929
Entrez Gene ID: (Human) 2027
Molecular Function: lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.