Novus Biologicals
Manufacturer Code:NBP216336
Catalog # NBP216336
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human ENO3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-phospho-D-glycerate hydro-lyase; 2-phospho-D-glycerate hydrolyase; 2-phospho-D-glycerate hydrolyase 2-phospho-D-glycerate hydro-lyase beta-enolase EC 4.2.1 EC 4.2.1.11 Enolase 3 enolase 3 (beta muscle) enolase 3 (beta muscle) GSD13 MSE Muscle-specific enolase Skeletal muscle enolase; Beta-enolase; Enolase 3; enolase 3 (beta, muscle); MSE; muscle enriched enolase; Muscle-specific enolase; Skeletal muscle enolase
Gene Aliases: ENO3; GSD13; MSE
UniProt ID: (Human) P13929
Entrez Gene ID: (Human) 2027
Molecular Function: lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.