Proteintech
Manufacturer Code:164211AP150UL
Catalog # 50-173-0934
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ENO3 Fusion Protein Ag9613 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-phospho-D-glycerate hydro-lyase; 2-phospho-D-glycerate hydrolyase; Beta enolase ENO3 Enolase 3 enolase 3 (beta muscle) MSE Muscle specific enolase Skeletal muscle enolase; Beta-enolase; Enolase 3; enolase 3 (beta, muscle); MSE; muscle enriched enolase; Muscle specific enolase (beta beta enolose) (two splicing products); Muscle-specific enolase; Skeletal muscle enolase
Gene Aliases: BBE; Eno-3; ENO3; GSD13; MSE
UniProt ID: (Human) P13929, (Mouse) P21550, (Rat) P15429
Entrez Gene ID: (Human) 2027, (Mouse) 13808, (Rat) 25438
Molecular Function: lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.