Invitrogen
Manufacturer Code:PA562080
Catalog # PIPA562080
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human EBPL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EBRP emopamil binding related protein delta8-delta7 sterol isomerase related protein emopamil-binding protein-like emopamil-binding-related protein; emopamil binding protein-like; emopamil binding related protein, delta8-delta7 sterol isomerase related protein; Emopamil-binding protein-like; Emopamil-binding-related protein
Gene Aliases: EBPL; EBRP; ERP
UniProt ID: (Human) Q9BY08
Entrez Gene ID: (Human) 84650
Molecular Function: isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.