Invitrogen
Manufacturer Code:MA191856
Catalog # PIMA191856
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 0.T.178 |
Immunogen | Recombit full length protein (Human). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: B-dynamin; D100; DNM; Dynamin, brain; Dynamin-1
Gene Aliases: AI838169; DNM; DNM1; EIEE31; Ftfl; Kiaa4093; mKIAA4093
UniProt ID: (Bovine) Q08DF4, (Human) Q05193, (Mouse) P39053, (Rat) P21575
Entrez Gene ID: (Bovine) 508794, (Human) 1759, (Mouse) 13429, (Rat) 140694
Molecular Function: G-protein cytoskeletal protein enzyme modulator hydrolase microtubule family cytoskeletal protein small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.