Invitrogen
Manufacturer Code:PA5109618
Catalog # PIPA5109618
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 150-250 of human DBH (NP_000778.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BOS_12212 Dbh DBM Dopamine beta-hydroxylase Dopamine beta-monooxygenase Dopamine-Beta-hydroxylase DOPBHY Soluble dopamine beta-hydroxylase; Dopamine beta hydroxylase (dopamine beta-monooxygenase); Dopamine beta-hydroxylase; dopamine beta-hydroxylase (dopamine beta-monooxygenase); Dopamine beta-monooxygenase; Soluble dopamine beta-hydroxylase
Gene Aliases: DBH; DBM; DOPBHY
UniProt ID: (Human) P09172, (Rat) Q05754, (Mouse) Q64237
Entrez Gene ID: (Human) 1621, (Rat) 25699, (Mouse) 13166
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.