Invitrogen
Manufacturer Code:PA5106498
Catalog # PIPA5106498
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DYR1A corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310043O08Rik D16Ertd272e D16Ertd493e dual specificity tyrosine phosphorylation regulated kinase 1A dual specificity tyrosine-(Y)-phosphorylation regulated kinase 1A Dual specificity tyrosine-phosphorylation-regulated kinase 1A Dual Specificity Yak1-related kinase dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1A DYRK DYRK1 DYRK1A ENSMUSG00000074897 Gm10783 hMNB HP86 mmb MNB mnb protein kinase homolog hp86 MNB/DYRK protein kinase MNBH Mp86 MRD7 OTTHUMP00000109090 protein kinase minibrain homolog PSK47 RP86 serine/threonine kinase MNB serine/threonine-specific protein kinase; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 1A; Dual specificity tyrosine-phosphorylation-regulated kinase 1A; Dual Specificity Yak1-related kinase; hMNB; HP86; mnb protein kinase homolog hp86; MNB/DYRK protein kinase; MNBH; MP86; Protein kinase minibrain homolog; RP86; serine/threonine kinase MNB; serine/threonine-specific protein kinase
Gene Aliases: 2310043O08Rik; D16Ertd272e; D16Ertd493e; DYRK; DYRK1; DYRK1A; ENSMUSG00000074897; Gm10783; HP86; mmb; MNB; MNBH; Mp86; MRD7; PSK47
UniProt ID: (Human) Q13627, (Mouse) Q61214, (Rat) Q63470
Entrez Gene ID: (Human) 1859, (Mouse) 13548, (Rat) 25255
Molecular Function: kinase non-receptor serine/threonine protein kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.