Abnova
Manufacturer Code:PAB2068
Catalog # 89-122-181
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to internal region of human DYRK1A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of DYRK1A.
For Research Use Only
Protein Aliases: dual specificity tyrosine-(Y)-phosphorylation regulated kinase 1A; Dual specificity tyrosine-phosphorylation-regulated kinase 1A; Dual specificity YAK1-related kinase; DYRK; DYRK1; hMNB; HP86; MNB; mnb protein kinase homolog hp86; MNB/DYRK protein kinase; MNBH; Protein kinase minibrain homolog; serine/threonine kinase MNB; serine/threonine-specific protein kinase
Gene Aliases: DYRK; DYRK1; DYRK1A; HP86; MNB; MNBH; MRD7
UniProt ID: (Human) Q13627
Entrez Gene ID: (Human) 1859
Molecular Function: kinase non-receptor serine/threonine protein kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.