Novus Biologicals
Manufacturer Code:NBP299035AF532
Catalog # NB057466
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant DR6/TNFRSF21 (NP_055267.1 Met1-Leu350). |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BM-018 CD358 antigen Death receptor 6 DR6TNFR-related death receptor 6 MGC31965 tumor necrosis factor receptor superfamily member 21 tumor necrosis factor receptor superfamily member 21; CD358; Death receptor 6; TNFR-related death receptor 6; Tumor necrosis factor receptor superfamily member 21; tumor necrosis factor receptor superfamily, member 21
Gene Aliases: BM-018; CD358; DR6; TNFRSF21; UNQ437/PRO868
UniProt ID: (Human) O75509
Entrez Gene ID: (Human) 27242
Molecular Function: cytokine receptor receptor tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.