Novus Biologicals
Manufacturer Code:NBP270583JF646
Catalog # N270583J646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7G2 |
Immunogen | Recombinant protein fragment corresponding to amino acids 1544-1830 of human DOCK2 (NP_004937) produced in E.coli. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: dedicator of cyto-kinesis 2; dedicator of cytokinesis 2 dedicator of cytokinesis protein 2 FLJ46592 KIAA0209dedicator of cyto-kinesis 2; Dedicator of cytokinesis protein 2
Gene Aliases: DOCK2; IMD40; KIAA0209
UniProt ID: (Human) Q92608
Entrez Gene ID: (Human) 1794
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.