Invitrogen
Manufacturer Code:PA543763
Catalog # PIPA543763
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human DOCK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dedicator of cyto-kinesis 2; Dedicator of cytokinesis protein 2; Protein Hch
Gene Aliases: AI662014; AW122239; CED-5; DOCK2; Hch; IMD40; KIAA0209; MBC
UniProt ID: (Human) Q92608, (Mouse) Q8C3J5
Entrez Gene ID: (Rabbit) 100357300, (Human) 1794, (Dog) 489128, (Guinea pig) 100714041, (Rat) 360509, (Mouse) 94176
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.